• USA Home
  • Search Results

  • 4methylheptane1142358953711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Methylheptane

1 Products
Image of 4-Methylheptane
CAS Number:
Linear Formula:
 (CH3CH2CH2)2CHCH3
Molecular Weight:
 114.23
 Beilstein/REAXYS Number:
 1696874
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
111023 4-Methylheptane, 99%,