• USA Home
  • Search Results

  • 4methylmphenylenediamine122179580711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Methyl-m-phenylenediamine

1 Products
Synonym:   2,4-Diaminotoluene, 2,4-Toluenediamine, 4-Methyl-1,3-phenylenediamine
Image of 4-Methyl-<I>m</I>-phenylenediamine
CAS Number:
Linear Formula:
 CH3C6H3(NH2)2
Molecular Weight:
 122.17
 Beilstein Registry Number:
 2205839
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More