• USA Home
  • Search Results

  • 4methylpyrimidine9411343846811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Methylpyrimidine

1 Products
Image of 4-Methylpyrimidine
CAS Number:
Empirical Formula (Hill Notation):
 C5H6N2
Molecular Weight:
 94.11
 Beilstein/REAXYS Number:
 105781
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
127493 4-Methylpyrimidine, 97%,