• USA Home
  • Search Results

  • 4methylumbelliferylphosphate25615336804511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Methylumbelliferyl phosphate

1 Products
Synonym:   4-Methylumbelliferyl-phosphoric acid
Image of 4-Methylumbelliferyl phosphate
CAS Number:
Empirical Formula (Hill Notation):
 C10H9O6P
Molecular Weight:
 256.15
 Beilstein/REAXYS Number:
 1687229
 EC Number:
 MDL number:
 NACRES:
 NA.32
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Fluorescence Test
Show More