• USA Home
  • Search Results

  • 4nitrobenzaldehydeoxime16613112937911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Nitrobenzaldehyde oxime

1 Products
Synonym:   4-Nitrobenzaldoxime
Image of 4-Nitrobenzaldehyde oxime
CAS Number:
Linear Formula:
 O2NC6H4CH=NOH
Molecular Weight:
 166.13
 Beilstein/REAXYS Number:
 2047086
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More