• USA Home
  • Search Results

  • 4nitrobenzylalcohol1531461973811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Nitrobenzyl alcohol

1 Products
Image of 4-Nitrobenzyl alcohol
CAS Number:
Linear Formula:
 O2NC6H4CH2OH
Molecular Weight:
 153.14
 Beilstein/REAXYS Number:
 1424026
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More