• USA Home
  • Search Results

  • 4nitrophenol1391110002711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Nitrophenol

6 Products
Synonym:   p-Nitrophenol
Image of 4-Nitrophenol
CAS Number:
Linear Formula:
 O2NC6H4OH
Molecular Weight:
 139.11
 Beilstein/REAXYS Number:
 1281877
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
 eCl@ss:
 39032018
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Anal. Standard Form
  Shelf Life
  Titertest
  Assay
  Physical Form
  Special Grade
Show More