• USA Home
  • Search Results

  • 4nitrophenylacetate1811583003511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Nitrophenyl acetate

3 Products
Synonym:   Acetic acid 4-nitrophenyl ester
Image of 4-Nitrophenyl acetate
CAS Number:
Linear Formula:
 CH3CO2C6H4NO2
Molecular Weight:
 181.15
 Beilstein/REAXYS Number:
 515874
 EC Number:
 MDL number:
 NACRES:
 NA.32
 PubChem Substance ID:
Popular Documents:  Product Information Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Melting Point
Show More