• USA Home
  • Search Results

  • 4nitrophenylbdglucuronide315231034494211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Nitrophenyl β-D-glucuronide

2 Products
Synonym:   4-Nitrophenyl-β-D-glucopyranosiduronic acid, PNPG
Image of 4-Nitrophenyl β-<SC>D</SC>-glucuronide
CAS Number:
Empirical Formula (Hill Notation):
 C12H13NO9
Molecular Weight:
 315.23
 Beilstein/REAXYS Number:
 1438576
 EC Number:
 MDL number:
 NACRES:
 NA.52
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More