• USA Home
  • Search Results

  • 4nitrophenylisocyanate1641210028711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Nitrophenyl isocyanate

1 Products
Image of 4-Nitrophenyl isocyanate
CAS Number:
Linear Formula:
 O2NC6H4NCO
Molecular Weight:
 164.12
 Beilstein/REAXYS Number:
 389516
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More