• USA Home
  • Search Results

  • 4nitrophenylnacetylbdgalactosaminide342301494896011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Nitrophenyl N-acetyl-β-D-galactosaminide

1 Products
Synonym:   4-Nitrophenyl 2-acetamido-2-deoxy-β-D-galactopyranoside, p-Nitrophenyl N-acetyl-β-D-galactosaminide
Image of 4-Nitrophenyl N-acetyl-β-<SC>D</SC>-galactosaminide
CAS Number:
Empirical Formula (Hill Notation):
 C14H18N2O8
Molecular Weight:
 342.30
 Beilstein/REAXYS Number:
 96194
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More