• USA Home
  • Search Results

  • 4nitrophenylphosphatedisodiumsalthexahydrate3711433333818411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Nitrophenyl phosphate disodium salt hexahydrate

5 Products
Synonym:   p-Nitrophenyl phosphate disodium hexahydrate, Disodium 4-nitrophenyl phosphate hexahydrate, di-Sodium 4-nitrophenyl phosphate, pNPP disodium hexahydrate
Image of 4-Nitrophenyl phosphate disodium salt hexahydrate
CAS Number:
Linear Formula:
 O2NC6H4OP(O)(ONa)2 · 6H2O
Molecular Weight:
 371.14
 Beilstein/REAXYS Number:
 5324661
 EC Number:
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Solubility
  Fluorescence Test
  Assay
Show More