• USA Home
  • Search Results

  • 4nitroquinolinenoxide190165657511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Nitroquinoline N-oxide

1 Products
Synonym:   4-NQO, 4-Nitroquinoline 1-oxide
Image of 4-Nitroquinoline <I>N</I>-oxide
CAS Number:
Empirical Formula (Hill Notation):
 C9H6N2O3
Molecular Weight:
 190.16
 Beilstein/REAXYS Number:
 165756
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More