• USA Home
  • Search Results

  • 4nmaleimidomethylcyclohexane1carboxylicacid3sulfonhydroxysuccinimideestersodiumsalt436379292124911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid 3-sulfo-N-hydroxysuccinimide ester sodium salt

1 Products
Synonym:   Sulfo-SMCC
Image of 4-(<I>N</I>-<WBR>Maleimidomethyl)<WBR>cyclohexane-<WBR>1-<WBR>carboxylic acid 3-sulfo-<I>N</I>-hydroxysuccinimide ester sodium salt
CAS Number:
Empirical Formula (Hill Notation):
 C16H17N2NaO9S
Molecular Weight:
 436.37
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Application(s)
Show More