• USA Home
  • Search Results

  • 4octyne11020194245611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Octyne

1 Products
Image of 4-Octyne
CAS Number:
Linear Formula:
 CH3CH2CH2C≡CCH2CH2CH3
Molecular Weight:
 110.20
 Beilstein/REAXYS Number:
 1732138
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
 eCl@ss:
 39010412
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
244473 4-Octyne, 99%,