• USA Home
  • Search Results

  • 4pacetoxyphenyl2butanone20624357206311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-(p-Acetoxyphenyl)-2-butanone

1 Products
Synonym:   4-(3-Oxobutyl)phenyl acetate, 4-(4-Acetoxyphenyl)-2-butanone, raspberry ketone acetate
Image of 4-(<I>p</I>-<WBR>Acetoxyphenyl)<WBR>-<WBR>2-<WBR>butanone
CAS Number:
Linear Formula:
 CH3CO2C6H4CH2CH2COCH3
Molecular Weight:
 206.24
 Beilstein/REAXYS Number:
 1961620
 EC Number:
 FEMA Number:
 3652
 Flavis number:
 9.288
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More