• USA Home
  • Search Results

  • 4phenylbenzylchloride20268166711411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Phenylbenzyl chloride

1 Products
Synonym:   4-(Chloromethyl)biphenyl
Image of 4-Phenylbenzyl chloride
CAS Number:
Linear Formula:
 C6H5C6H4CH2Cl
Molecular Weight:
 202.68
 Beilstein/REAXYS Number:
 1863327
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More