• USA Home
  • Search Results

  • 4tertamylphenol164248046611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-tert-Amylphenol

2 Products
Image of 4-<I>tert</I>-Amylphenol
CAS Number:
Linear Formula:
 C2H5C(CH3)2C6H4OH
Molecular Weight:
 164.24
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Application(s)
  Assay
Show More