• USA Home
  • Search Results

  • 4tertbutylcatechol166229829311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-tert-Butylcatechol

2 Products
Image of 4-<I>tert</I>-Butylcatechol
CAS Number:
Linear Formula:
 (CH3)3CC6H3-1,2-(OH)2
Molecular Weight:
 166.22
 Beilstein/REAXYS Number:
 2043335
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Solubility
  Melting Point
Show More