• USA Home
  • Search Results

  • 4vinyl1cyclohexene1081810040311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

4-Vinyl-1-cyclohexene

1 Products
Synonym:   4-Ethenyl-1-cyclohexene, NSC 15760
Image of 4-<WBR>Vinyl-<WBR>1-<WBR>cyclohexene
CAS Number:
Linear Formula:
 C6H9CH=CH2
Molecular Weight:
 108.18
 Beilstein/REAXYS Number:
 1901553
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Titertest
Show More