• USA Home
  • Search Results

  • 5101520tetra4pyridyl21h23hporphine618691683413211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5,10,15,20-Tetra(4-pyridyl)-21H,23H-porphine

1 Products
Synonym:   5,10,15,20-Tetra(4-pyridyl)porphyrin
Image of 5,10,15,20-<WBR>Tetra(4-<WBR>pyridyl)<WBR>-<WBR>21<I>H</I>,23<I>H</I>-porphine
CAS Number:
Empirical Formula (Hill Notation):
 C40H26N8
Molecular Weight:
 618.69
 Beilstein/REAXYS Number:
 635257
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More