• USA Home
  • Search Results

  • 5101520tetrakis4methoxyphenyl21h23hporphinecobaltii791762890371111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphine cobalt(II)

1 Products
Image of 5,10,15,20-<WBR>Tetrakis(4-<WBR>methoxyphenyl)<WBR>-<WBR>21<I>H</I>,23<I>H</I>-porphine cobalt(II)
CAS Number:
Empirical Formula (Hill Notation):
 C48H36CoN4O4
Molecular Weight:
 791.76
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Absorption
Show More