• USA Home
  • Search Results

  • 5101520tetrakispentafluorophenylporphyrin974552544014611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5,10,15,20-Tetrakis(pentafluorophenyl)porphyrin

1 Products
Image of 5,10,15,20-<WBR>Tetrakis(pentafluorophenyl)<WBR>porphyrin
CAS Number:
Empirical Formula (Hill Notation):
 C44H10F20N4
Molecular Weight:
 974.55
 Beilstein/REAXYS Number:
 604408
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Absorption
Show More