• USA Home
  • Search Results

  • 5101520tetraphenyl21h23hporphine6147491723711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5,10,15,20-Tetraphenyl-21H,23H-porphine

2 Products
Synonym:   meso-Tetraphenylporphyrin, TPP
Image of 5,10,15,20-<WBR>Tetraphenyl-<WBR>21<I>H</I>,23<I>H</I>-porphine
CAS Number:
Empirical Formula (Hill Notation):
 C44H30N4
Molecular Weight:
 614.74
 Beilstein/REAXYS Number:
 379542
 EC Number:
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Physical Form
  Solubility
  Assay
  Absorption
Show More