• USA Home
  • Search Results

  • 5101520tetraphenyl21h23hporphinezinc678111407480711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5,10,15,20-Tetraphenyl-21H,23H-porphine zinc

2 Products
Synonym:   meso-Tetraphenylporphine, TPP, Zinc TPP
Image of 5,10,15,20-<WBR>Tetraphenyl-<WBR>21<I>H</I>,23<I>H</I>-porphine zinc
CAS Number:
Empirical Formula (Hill Notation):
 C44H28N4Zn
Molecular Weight:
 678.11
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Absorption
  OLED Device Performance
Show More