• USA Home
  • Search Results

  • 55dimethyl1pyrrolinenoxide11316331761111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5,5-Dimethyl-1-pyrroline N-oxide

2 Products
Synonym:   DMPO
Image of 5,5-<WBR>Dimethyl-<WBR>1-<WBR>pyrroline <I>N</I>-oxide
CAS Number:
Empirical Formula (Hill Notation):
 C6H11NO
Molecular Weight:
 113.16
 Beilstein/REAXYS Number:
 107603
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More