• USA Home
  • Search Results

  • 57dichloro8hydroxy2methylquinoline228077280011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5,7-Dichloro-8-hydroxy-2-methylquinoline

1 Products
Synonym:   5,7-Dichloro-2-methyl-8-quinolinol, 5,7-Dichloro-8-hydroxyquinaldine, 5,7-Dichloro-8-quinaldinol, BCM
Image of 5,7-<WBR>Dichloro-<WBR>8-<WBR>hydroxy-<WBR>2-<WBR>methylquinoline
CAS Number:
Empirical Formula (Hill Notation):
 C10H7Cl2NO
Molecular Weight:
 228.07
 Beilstein/REAXYS Number:
 156683
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
Show More