• USA Home
  • Search Results

  • 57dihydroxy4methylcoumarin19217210776811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5,7-Dihydroxy-4-methylcoumarin

2 Products
Image of 5,7-<WBR>Dihydroxy-<WBR>4-<WBR>methylcoumarin
CAS Number:
Empirical Formula (Hill Notation):
 C10H8O4
Molecular Weight:
 192.17
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More