• USA Home
  • Search Results

  • 57dimethoxycoumarin2061948706911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5,7-Dimethoxycoumarin

1 Products
Synonym:   Citropten, Limettin
Image of 5,7-<WBR>Dimethoxycoumarin
CAS Number:
Empirical Formula (Hill Notation):
 C11H10O4
Molecular Weight:
 206.19
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More