• USA Home
  • Search Results

  • 5aandrost16en3aol27444115351111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5α-Androst-16-en-3α-ol

1 Products
Synonym:   16-(5α)Androsten-3α-ol, 3α-Hydroxy-5α-androst-16-ene, Androstenol
Image of 5α-Androst-16-en-3α-ol
CAS Number:
Empirical Formula (Hill Notation):
 C19H30O
Molecular Weight:
 274.44
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
A7883 5α-Androst-16-en-3α-ol,