• USA Home
  • Search Results

  • 5adihydrotestosteroned3161617d3solution293467903734611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5α-Dihydrotestosterone-D3 (16,16,17-D3) solution

1 Products
Synonym:   5α-DHT-D3 (16, 16, 17-D3) solution, 5α-Dihydrotestosterone-d3 (16,16,17-d3)
Image of 5α-Dihydrotestosterone-D<SUB>3</SUB> (16,16,17-D<SUB>3</SUB>) solution
CAS Number:
Empirical Formula (Hill Notation):
 C19H27O2D3
Molecular Weight:
 293.46
 EC Number:
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More