• USA Home
  • Search Results

  • 5amino4imidazolecarboxamide1261236097411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5-Amino-4-imidazolecarboxamide

1 Products
Image of 5-<WBR>Amino-<WBR>4-<WBR>imidazolecarboxamide
CAS Number:
Empirical Formula (Hill Notation):
 C4H6N4O
Molecular Weight:
 126.12
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More