• USA Home
  • Search Results

  • 5aminolevulinicacidhydrochloride16759545109211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5-Aminolevulinic acid hydrochloride

3 Products
Synonym:   δ-Aminolevulinic acid hydrochloride, 5-Amino-4-oxopentanoic acid hydrochloride, 5-Aminolaevulinic acid hydrochloride, ALA
Image of 5-Aminolevulinic acid hydrochloride
CAS Number:
Linear Formula:
 NH2CH2C(O)CH2CH2COOH · HCl
Molecular Weight:
 167.59
 Beilstein/REAXYS Number:
 3690651
 EC Number:
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Color
  Melting Point
  Physical Form
  Application(s)
Show More