• USA Home
  • Search Results

  • 5aza2deoxycytidine22821235333511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5-Aza-2′-deoxycytidine

1 Products
Synonym:   2′-Deoxy-5-azacytidine, 4-Amino-1-(2-deoxy-β-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one, Decitabine
Image of 5-Aza-2′-deoxycytidine
CAS Number:
Empirical Formula (Hill Notation):
 C8H12N4O4
Molecular Weight:
 228.21
 Beilstein/REAXYS Number:
 617982
 EC Number:
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More