• USA Home
  • Search Results

  • 5bromo123trifluorobenzene2109813852669911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5-Bromo-1,2,3-trifluorobenzene

1 Products
Synonym:   1-Bromo-3,4,5-trifluorobenzene
Image of 5-<WBR>Bromo-<WBR>1,2,3-<WBR>trifluorobenzene
CAS Number:
Linear Formula:
 BrC6H2F3
Molecular Weight:
 210.98
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More