• USA Home
  • Search Results

  • 5bromo2deoxyuridine307105914311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5-Bromo-2′-deoxyuridine

3 Products
Synonym:   5-BrdU, 5-Bromo-1-(2-deoxy-β-D-ribofuranosyl)uracil, 5-Bromouracil deoxyriboside, BUdR
Image of 5-Bromo-2′-deoxyuridine
CAS Number:
Empirical Formula (Hill Notation):
 C9H11BrN2O5
Molecular Weight:
 307.10
 Beilstein/REAXYS Number:
 30395
 EC Number:
 MDL number:
 NACRES:
 NA.51
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More