• USA Home
  • Search Results

  • 5bromo4chloro3indolylbdgalactopyranoside40863724090611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5-Bromo-4-chloro-3-indolyl β-D-galactopyranoside

3 Products
Synonym:   5-Bromo-4-chloro-3-indolyl β-D-galactoside, BCIG, X-Gal
Image of 5-<WBR>Bromo-<WBR>4-<WBR>chloro-<WBR>3-<WBR>indolyl β-<SC>D</SC>-galactopyranoside
CAS Number:
Empirical Formula (Hill Notation):
 C14H15BrClNO6
Molecular Weight:
 408.63
 Beilstein/REAXYS Number:
 1402009
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More