• USA Home
  • Search Results

  • 5bromo4chloro3indolylphosphateptoluidinesalt43362657806911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt

4 Products
Synonym:   BCIP® p-toluidine salt, X-phosphate p-toluidine salt
Image of 5-<WBR>Bromo-<WBR>4-<WBR>chloro-<WBR>3-<WBR>indolyl phosphate <I>p</I>-toluidine salt
CAS Number:
Empirical Formula (Hill Notation):
 C8H6BrClNO4P · C7H9N
Molecular Weight:
 433.62
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Product Information Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Solubility
Show More