• USA Home
  • Search Results

  • 5bromo6chloro3indolylbdglucuronidecyclohexylammoniumsalt5217914411043011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5-Bromo-6-chloro-3-indolyl β-D-glucuronide cyclohexylammonium salt

1 Products
Synonym:   Magenta-GlcA, Magenta glucuronide
Image of 5-<WBR>Bromo-<WBR>6-<WBR>chloro-<WBR>3-<WBR>indolyl β-<SC>D</SC>-glucuronide cyclohexylammonium salt
CAS Number:
Empirical Formula (Hill Notation):
 C14H13BrClNO7 · C6H13N
Molecular Weight:
 521.79
 MDL number:
 NACRES:
 NA.83
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More