• USA Home
  • Search Results

  • 5bromoindole196041007550011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5-Bromoindole

1 Products
Image of 5-Bromoindole
CAS Number:
Empirical Formula (Hill Notation):
 C8H6BrN
Molecular Weight:
 196.04
 Beilstein/REAXYS Number:
 112877
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
B68607 5-Bromoindole, 99%,