• USA Home
  • Search Results

  • 5carboxyfluorescein376327682303511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5-Carboxyfluorescein

3 Products
Synonym:   5-FAM
Image of 5-Carboxyfluorescein
CAS Number:
Empirical Formula (Hill Notation):
 C21H12O7
Molecular Weight:
 376.32
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Assay
Show More