• USA Home
  • Search Results

  • 5chloro8quinolinol1796013016511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5-Chloro-8-quinolinol

1 Products
Synonym:   5-Chloro-8-hydroxyquinoline
Image of 5-<WBR>Chloro-<WBR>8-<WBR>quinolinol
CAS Number:
Empirical Formula (Hill Notation):
 C9H6ClNO
Molecular Weight:
 179.60
 Beilstein/REAXYS Number:
 5289
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More