• USA Home
  • Search Results

  • 5chlorouracil14653182081111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5-Chlorouracil

1 Products
Synonym:   5-Chloro-2,4-dihydroxypyrimidine, 5-Chloropyrimidine-2,4(1H,3H)-dione
Image of 5-Chlorouracil
CAS Number:
Empirical Formula (Hill Notation):
 C4H3ClN2O2
Molecular Weight:
 146.53
 Beilstein/REAXYS Number:
 127173
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
Show More