• USA Home
  • Search Results

  • 5difluoromethoxy2mercapto1hbenzimidazole216219796362711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5-(Difluoromethoxy)-2-mercapto-1H-benzimidazole

1 Products
Image of 5-<WBR>(Difluoromethoxy)<WBR>-<WBR>2-<WBR>mercapto-<WBR>1<I>H</I>-benzimidazole
CAS Number:
Empirical Formula (Hill Notation):
 C8H6F2N2OS
Molecular Weight:
 216.21
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More