• USA Home
  • Search Results

  • 5fluoro2hydroxypyrimidine11408202278811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5-Fluoro-2-hydroxypyrimidine

1 Products
Synonym:   5-Fluoro-2(1H)-pyrimidone
Image of 5-<WBR>Fluoro-<WBR>2-<WBR>hydroxypyrimidine
CAS Number:
Empirical Formula (Hill Notation):
 C4H3FN2O
Molecular Weight:
 114.08
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More