• USA Home
  • Search Results

  • 5fluoro5deoxyuridine24619309409511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5-Fluoro-5′-deoxyuridine

1 Products
Synonym:   5′dFUrd, Doxifluridine
Image of 5-Fluoro-5′-deoxyuridine
CAS Number:
Empirical Formula (Hill Notation):
 C9H11FN2O5
Molecular Weight:
 246.19
 EC Number:
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
F8791 5-Fluoro-5′-deoxyuridine,