• USA Home
  • Search Results

  • 5fluorouracil130085121811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5-Fluorouracil

4 Products
Synonym:   2,4-Dihydroxy-5-fluoropyrimidine, 5-FU, 5-Fluoro-2,4(1H,3H)-pyrimidinedione
Image of 5-Fluorouracil
CAS Number:
Empirical Formula (Hill Notation):
 C4H3FN2O2
Molecular Weight:
 130.08
 Beilstein/REAXYS Number:
 127172
 EC Number:
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Shelf Life
  Special Grade
  Solubility
Show More