• USA Home
  • Search Results

  • 5hydroxyindole13315195354411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5-Hydroxyindole

1 Products
Synonym:   5-Hydroxyindole, 5-Indolol, NSC 87503
Image of 5-Hydroxyindole
CAS Number:
Empirical Formula (Hill Notation):
 C8H7NO
Molecular Weight:
 133.15
 Beilstein/REAXYS Number:
 112349
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
H31859 5-Hydroxyindole, 97%,