• USA Home
  • Search Results

  • 5hydroxyindole2carboxylicacid177162159806111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5-Hydroxyindole-2-carboxylic acid

1 Products
Synonym:   NSC 117338
Image of 5-<WBR>Hydroxyindole-<WBR>2-<WBR>carboxylic acid
CAS Number:
Empirical Formula (Hill Notation):
 C9H7NO3
Molecular Weight:
 177.16
 Beilstein/REAXYS Number:
 153214
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More