• USA Home
  • Search Results

  • 5iodo2deoxyuridine354105442211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

5-Iodo-2′-deoxyuridine

1 Products
Synonym:   1-(2-Deoxy-β-D-ribofuranosyl)-5-iodouracil, 2′-Deoxy-5-iodouridine, 5-IUdR, IDU, IdUrd, Idoxuridine
Image of 5-Iodo-2′-deoxyuridine
CAS Number:
Empirical Formula (Hill Notation):
 C9H11IN2O5
Molecular Weight:
 354.10
 Beilstein/REAXYS Number:
 30397
 EC Number:
 MDL number:
 NACRES:
 NA.51
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More